Novus Biologicals
Manufacturer Code:NBP203269
Catalog # NBP203269
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI1B3 |
Immunogen | Full length human recombinant protein of human HDAC6 (NP_006035) produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: EC 3.5.1.98 HD6FLJ16239 histone deacetylase 6 JM21 KIAA0901; HD6; Histone deacetylase 6; protein phosphatase 1, regulatory subunit 90; Tubulin-lysine deacetylase HDAC6
Gene Aliases: CPBHM; HD6; HDAC6; JM21; KIAA0901; PPP1R90
UniProt ID: (Human) Q9UBN7
Entrez Gene ID: (Human) 10013
Molecular Function:
deacetylase
hydrolase
nucleic acid binding
oxidoreductase
reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.