Invitrogen
Manufacturer Code:PA582914
Catalog # PIPA582914
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human HDAC6 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FLJ16239 HD6 JM21 KIAA0901 PPP1R90; HD6; Histone deacetylase 6; protein phosphatase 1, regulatory subunit 90; Tubulin-lysine deacetylase HDAC6
Gene Aliases: CPBHM; HD6; HDAC6; JM21; KIAA0901; PPP1R90
UniProt ID: (Human) Q9UBN7
Entrez Gene ID: (Human) 10013
Molecular Function: deacetylase hydrolase nucleic acid binding oxidoreductase reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.