Proteintech
Manufacturer Code:161671AP150UL
Catalog # 50-173-0766
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HD6; HD6 HDAC6 histone deacetylase 6 JM21 KIAA0901; histone deacetylase 5; Histone deacetylase 6; Histone deacetylase mHDA2; protein phosphatase 1, regulatory subunit 90; scurfy candidate 6; Tubulin-lysine deacetylase HDAC6
Gene Aliases: CPBHM; HD6; Hdac5; HDAC6; JM21; KIAA0901; mHDA2; PPP1R90; Sfc6
UniProt ID: (Human) Q9UBN7, (Mouse) Q9Z2V5
Entrez Gene ID: (Human) 10013, (Rat) 84581, (Mouse) 15185
Molecular Function:
deacetylase
hydrolase
nucleic acid binding
oxidoreductase
reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.