Invitrogen
Manufacturer Code:PA5105299
Catalog # PIPA5105299
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human HDAC5 corresponding to a region within the internal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AI426555 antigen NY-CO-9 DKFZp779K1053 FLJ90614 HD5 HD7 HD7b HD9 HDAC Hdac4 HDAC5 HDAC-5 HDAC7 HDAC7B HDAC9 HDAC9B HDAC9FL HDRP histone deacetylase 4 histone deacetylase 5 histone deacetylase mHDA1 KIAA0600 KIAA0744 mHDA1 mKIAA0600 NY-CO-9; Antigen NY-CO-9; HD5; histone deacetylase 4; Histone deacetylase 5; histone deacetylase mHDA1
Gene Aliases: AI426555; HD5; Hdac4; HDAC5; KIAA0600; mHDA1; mKIAA0600; NY-CO-9
UniProt ID: (Human) Q9UQL6
Entrez Gene ID: (Human) 10014, (Mouse) 15184
Molecular Function:
deacetylase
hydrolase
nucleic acid binding
oxidoreductase
reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.