Novus Biologicals
Manufacturer Code:NBP203989
Catalog # NBP203989
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was generated by immunizing rabbits with a synthetic non-phosphopeptide corresponding to human HDAC5 around the phosphorylation site of serine 498 (T-Q-SP-S-P). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Antigen NY-CO-9; Antigen NY-CO-9 EC 3.5.1.98 FLJ90614 HD5 histone deacetylase 5 KIAA0600 NY-CO-9; HD5; Histone deacetylase 5
Gene Aliases: HD5; HDAC5; KIAA0600; NY-CO-9
UniProt ID: (Human) Q9UQL6
Entrez Gene ID: (Human) 10014
Molecular Function:
deacetylase
hydrolase
nucleic acid binding
oxidoreductase
reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.