OriGene
Manufacturer Code:TA805058
Catalog # 50-168-0533
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI4D7 |
Immunogen | Full length human recombit protein of human HDAC4 produced in HEK293T cell. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: AHO3 BDMR HA6116 HD4 HDAC-4 HDAC-A HDACA; HD4; Histone deacetylase 4; histone deacetylase A
Gene Aliases: AHO3; BDMR; HA6116; HD4; HDAC-4; HDAC-A; HDAC4; HDACA; KIAA0288
UniProt ID: (Human) P56524
Entrez Gene ID: (Human) 9759
Molecular Function: deacetylase hydrolase nucleic acid binding oxidoreductase reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.