Proteintech
Manufacturer Code:161651AP150UL
Catalog # 50-173-0764
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HA6116 HD4 HDAC A HDAC4 HDACA histone deacetylase 4 KIAA0288; HD4; Histone deacetylase 4; histone deacetylase A
Gene Aliases: 4932408F19Rik; AHO3; AI047285; BDMR; HA6116; HD4; HDAC-4; HDAC-A; HDAC4; HDACA; KIAA0288
UniProt ID: (Human) P56524, (Rat) Q99P99, (Mouse) Q6NZM9
Entrez Gene ID: (Human) 9759, (Rat) 363287, (Mouse) 208727
Molecular Function:
deacetylase
hydrolase
nucleic acid binding
oxidoreductase
reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.