OriGene
Manufacturer Code:TA808327
Catalog # 50-168-2056
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI7E10 |
Immunogen | Human recombit protein fragment corresponding to amino acids 415-488 of human HDAC2 produced in E.coli. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: HD2; HD2 RPD3 YAF1; Histone deacetylase 2; transcriptional regulator homolog RPD3; YY1-associated factor 1
Gene Aliases: HD2; HDAC2; RPD3; YAF1
UniProt ID: (Human) Q92769
Entrez Gene ID: (Human) 3066
Molecular Function:
deacetylase
hydrolase
nucleic acid binding
oxidoreductase
reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.