Novus Biologicals
Manufacturer Code:NB10056340SS
Catalog # NB10056340T
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was generated by immunizing rabbits with a mixture of synthetic peptides corresponding to amino acids 1-5 433-448 and 467-482 of human HDAC1 (Genbank Accession no. Q13547). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 3.5.1.98 GON-10 HD1DKFZp686H12203 histone deacetylase 1 reduced potassium dependency yeast homolog-like 1 RPD3L1RPD3; HD1; Histone deacetylase 1; reduced potassium dependency, yeast homolog-like 1
Gene Aliases: GON-10; HD1; HDAC1; RPD3; RPD3L1
UniProt ID: (Human) Q13547
Entrez Gene ID: (Human) 3065
Molecular Function: deacetylase hydrolase nucleic acid binding oxidoreductase reductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.