Novus Biologicals
Manufacturer Code:NBP212910AF647
Catalog # NBP212910Y
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S114-10 |
Immunogen | Fusion protein amino acids 1-019-1198 (c-terminus) of rat HCN4 |
Conjugate | Alexa Fluor 647 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C in the dark. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: hyperpolarization activated cyclic nucleotide-gated cation channel 4; hyperpolarization activated cyclic nucleotide-gated cation channel 4 hyperpolarization activated cyclic nucleotide-gated potassium channel 4 potassium/sodium hyperpolarization-activated cyclic nucleotide-gated channel 4 SSS2; hyperpolarization activated cyclic nucleotide-gated potassium channel 4; Potassium/sodium hyperpolarization-activated cyclic nucleotide-gated channel 4
Gene Aliases: HCN4; SSS2
UniProt ID: (Human) Q9Y3Q4
Entrez Gene ID: (Human) 10021
Molecular Function: cyclic nucleotide-gated ion channel ion channel ligand-gated ion channel potassium channel receptor transporter voltage-gated ion channel voltage-gated potassium channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.