Invitrogen
Manufacturer Code:PA5111878
Catalog # PIPA5111878
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | GST fusion protein corresponding to amino acid residues 119-155 of human HCN4 (Intracellular N-terminus) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BCNG-3; Bcng3 Hcn3 Hcn4 Potassium/sodium hyperpolarization-activated cyclic nucleotide-gated channel 4 SSS2; brain cyclic nucleotide-gated channel 3; hyperpolarization activated cyclic nucleotide-gated cation channel 4; hyperpolarization activated cyclic nucleotide-gated potassium channel 4; hyperpolarization-activated, cyclic nucleotide-gated K+ 3; hyperpolarization-activated, cyclic nucleotide-gated K+ 4; Potassium/sodium hyperpolarization-activated cyclic nucleotide-gated channel 4
Gene Aliases: Bcng3; Hcn3; HCN4; SSS2
UniProt ID: (Human) Q9Y3Q4, (Rat) Q9JKA7
Entrez Gene ID: (Human) 10021, (Mouse) 330953, (Rat) 59266
Molecular Function: cyclic nucleotide-gated ion channel ion channel ligand-gated ion channel potassium channel receptor transporter voltage-gated ion channel voltage-gated potassium channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.