Novus Biologicals
Manufacturer Code:NBP212895
Catalog # NBP212895
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Frozen) (IHC (F)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S71-37 |
Immunogen | Fusion protein amino acids 761-863 of rat HCN2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: BCNG-2; BCNG2 BCNG-2 brain cyclic nucleotide gated channel 2 brain cyclic nucleotide-gated channel 2 HAC-1 hyperpolarization activated cyclic nucleotide-gated potassium channel 2 potassium/sodium hyperpolarization-activated cyclic nucleotide-gated channel 2; brain cyclic nucleotide gated channel 2; Brain cyclic nucleotide-gated channel 2; hyperpolarization activated cyclic nucleotide-gated potassium channel 2; Potassium/sodium hyperpolarization-activated cyclic nucleotide-gated channel 2
Gene Aliases: BCNG-2; BCNG2; HAC-1; HCN2
UniProt ID: (Human) Q9UL51
Entrez Gene ID: (Human) 610
Molecular Function: cyclic nucleotide-gated ion channel ion channel ligand-gated ion channel potassium channel receptor transporter voltage-gated ion channel voltage-gated potassium channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.