Invitrogen
Manufacturer Code:PA577594
Catalog # PIPA577594
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide (C)EEAGPAGEPRGSQAS corresponding to amino acid residues 147-161 of human HCN2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: BCNG-2; brain cyclic nucleotide gated channel 2; Brain cyclic nucleotide-gated channel 2; HAC-1; hyperpolarization activated cyclic nucleotide-gated potassium channel 2; Hyperpolarization-activated cation channel 1; Hyperpolarization-Activated Cyclic Nucleotide-Gated Cation Channel 2 BCNG2 HAC-1; Potassium/sodium hyperpolarization-activated cyclic nucleotide-gated channel 2
Gene Aliases: BCNG-2; BCNG2; HAC-1; HAC1; HCN2; trls
UniProt ID: (Human) Q9UL51, (Mouse) O88703, (Rat) Q9JKA9
Entrez Gene ID: (Human) 610, (Mouse) 15166, (Rat) 114244
Molecular Function:
cyclic nucleotide-gated ion channel
ion channel
ligand-gated ion channel
potassium channel
receptor
transporter
voltage-gated ion channel
voltage-gated potassium channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.