Novus Biologicals
Manufacturer Code:NBP122450
Catalog # NBP122450
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | S70-28 |
Immunogen | Fusion protein amino acids 778-910 (C terminus) of rat HCN1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: BCNG-1; BCNG1 BCNG-1brain cyclic nucleotide gated channel 1 Brain cyclic nucleotide-gated channel 1 HAC-2 hyperpolarization activated cyclic nucleotide-gated potassium channel 1 potassium/sodium hyperpolarization-activated cyclic nucleotide-gated channel 1; Brain cyclic nucleotide-gated channel 1; Potassium/sodium hyperpolarization-activated cyclic nucleotide-gated channel 1
Gene Aliases: BCNG-1; BCNG1; EIEE24; HAC-2; HCN1
UniProt ID: (Human) O60741
Entrez Gene ID: (Human) 348980
Molecular Function:
cyclic nucleotide-gated ion channel
ion channel
ligand-gated ion channel
potassium channel
receptor
transporter
voltage-gated ion channel
voltage-gated potassium channel
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.