Invitrogen
Manufacturer Code:MA537681
Catalog # PIMA537681
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1508CT602.13.1 |
Immunogen | recombinant protein of human HCK |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: AI849071 B cell/myeloid kinase B-cell/myeloid kinase BMK HCK HCK proto-oncogene Src family tyrosine kinase Hctk Hematopoietic cell kinase hemopoietic cell kinase hemopoietic cell tyrosine kinase JTK9 p56Hck p56-HCK p56-HCK/p59-HCK p59Hck p59-HCK/p60-HCK p61Hck Tyrosine-protein kinase HCK; Hematopoietic cell kinase; Hemopoietic cell kinase; p59-HCK/p60-HCK; p59Hck; p61Hck; Tyrosine-protein kinase HCK
Gene Aliases: HCK; JTK9; p59Hck; p61Hck
UniProt ID: (Human) P08631
Entrez Gene ID: (Human) 3055
Molecular Function:
kinase
non-receptor tyrosine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.