Invitrogen
Manufacturer Code:PA5109806
Catalog # PIPA5109806
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-220 of human HCK (NP_001165600.1). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AI849071 B-cell/myeloid kinase BMK HCK Hctk Hematopoietic cell kinase Hemopoietic cell kinase JTK9 p56Hck p56-HCK p56-HCK/p59-HCK p59Hck p59-HCK/p60-HCK p61Hck Tyrosine-protein kinase HCK; Hematopoietic cell kinase; Hemopoietic cell kinase; p59-HCK/p60-HCK; p59Hck; p61Hck; Tyrosine-protein kinase HCK
Gene Aliases: HCK; JTK9; p59Hck; p61Hck
UniProt ID: (Human) P08631
Entrez Gene ID: (Human) 3055
Molecular Function:
kinase
non-receptor tyrosine protein kinase
protein kinase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.