Invitrogen
Manufacturer Code:PA5105729
Catalog # PIPA5105729
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human HCK corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AI849071 B cell/myeloid kinase B-cell/myeloid kinase BMK HCK HCK proto-oncogene Src family tyrosine kinase Hctk Hematopoietic cell kinase hemopoietic cell kinase hemopoietic cell tyrosine kinase JTK9 p56Hck p56-HCK p56-HCK/p59-HCK p59Hck p59-HCK/p60-HCK p61Hck Tyrosine-protein kinase HCK; B cell/myeloid kinase; B-cell/myeloid kinase; BMK; Hematopoietic cell kinase; Hemopoietic cell kinase; p56-HCK/p59-HCK; p59-HCK/p60-HCK; p59Hck; p61Hck; Tyrosine-protein kinase HCK
Gene Aliases: AI849071; Bmk; HCK; JTK9; p59Hck; p61Hck
UniProt ID: (Human) P08631, (Mouse) P08103
Entrez Gene ID: (Human) 3055, (Mouse) 15162
Molecular Function: kinase non-receptor tyrosine protein kinase protein kinase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.