Invitrogen
Manufacturer Code:PA5102495
Catalog # PIPA5102495
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human HCFC1 corresponding to a region within N-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AW060991 C1 factor CFF HCF HCF C-terminal chain 1 HCF C-terminal chain 2 HCF C-terminal chain 3 HCF C-terminal chain 4 HCF C-terminal chain 5 HCF C-terminal chain 6 HCF N-terminal chain 1 HCF N-terminal chain 2 HCF N-terminal chain 3 HCF N-terminal chain 4 HCF N-terminal chain 5 HCF N-terminal chain 6 HCF1 HCF-1 Hcfc1 HFC1 Host cell factor 1 host cell factor C1 MRX3 PPP1R89 protein phosphatase 1 regulatory subunit 89 RGD1563804 VCAF VP16 accessory protein VP16-accessory protein; C1 factor; CFF; HCF; HCF C-terminal chain 1; HCF C-terminal chain 2; HCF C-terminal chain 3; HCF C-terminal chain 4; HCF C-terminal chain 5; HCF C-terminal chain 6; HCF N-terminal chain 1; HCF N-terminal chain 2; HCF N-terminal chain 3; HCF N-terminal chain 4; HCF N-terminal chain 5; HCF N-terminal chain 6; hcfc1; Host cell factor 1; protein phosphatase 1, regulatory subunit 89; VCAF; VP16 accessory protein; VP16-accessory protein
Gene Aliases: AW060991; CFF; HCF; HCF-1; HCF1; HCFC1; HFC1; MRX3; PPP1R89; RGD1563804; VCAF
UniProt ID: (Human) P51610, (Mouse) Q9QWH2
Entrez Gene ID: (Human) 3054, (Rat) 363519, (Mouse) 15161
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.