Invitrogen
Manufacturer Code:PA5110311
Catalog # PIPA5110311
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-260 of human RNGTT (NP_003791.3). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: AU020997 Cap1a GTase GTP--RNA guanylyltransferase hCAP HCAP1 HCE HCE1 MCE1 mRNA 5&prime-triphosphatase mRNA guanylyltransferase mRNA-capping enzyme Polynucleotide 5&prime-triphosphatase RNGTT RP3-488C13.1 TPase; GTase; GTP--RNA guanylyltransferase; HCAP1; HCE; MCE1; mRNA 5'-triphosphatase; mRNA guanylyltransferase; mRNA-capping enzyme; Polynucleotide 5'-triphosphatase; TPase
Gene Aliases: AU020997; CAP1A; hCAP; HCE; HCE1; MCE1; RNGTT
UniProt ID: (Human) O60942, (Mouse) O55236
Entrez Gene ID: (Human) 8732, (Mouse) 24018, (Rat) 313131
Molecular Function:
hydrolase
nucleotidyltransferase
phosphatase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.