Invitrogen
Manufacturer Code:PA597824
Catalog # PIPA597824
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit Human Cytochrome c-type heme lyase protein (1-268AA) |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CCHL; cytochrome c heme-lyase; Cytochrome c-type heme lyase; Holocytochrome c-type synthase; microphthalamia with linear skin defects
Gene Aliases: CCHL; HCCS; LSDMCA1; MCOPS7; MLS
UniProt ID: (Human) P53701, (Mouse) P53702
Entrez Gene ID: (Human) 3052, (Mouse) 15159
Molecular Function: lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.