Novus Biologicals
Manufacturer Code:NBP186577
Catalog # NBP186577
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | This antibody was developed against Recombinant Protein corresponding to amino acids:YVECPIRGTAAENKENLDPSNLMPPPNQTPAPDQPFALSTVREESSIPRADSEKKWVYPSEQMFWNAMLKKGWKWKDEDISQKDMYNIIRIHNQNNEQAWKEILKWEALHAAECPCGPSLIRFGGKAKEYSPRARIRSWMGYEL |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CCHLDKFZp779I1858 cytochrome c heme-lyase cytochrome c-type heme lyase EC 4.4.1.17 holocytochrome c synthase holocytochrome c synthase (cytochrome c heme-lyase) Holocytochrome c-type synthase MCOPS7; cytochrome c heme-lyase; Cytochrome c-type heme lyase; Holocytochrome c-type synthase; microphthalamia with linear skin defects
Gene Aliases: CCHL; HCCS; LSDMCA1; MCOPS7; MLS
UniProt ID: (Human) P53701
Entrez Gene ID: (Human) 3052
Molecular Function: lyase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.