Novus Biologicals
Manufacturer Code:NBP30485620UL
Catalog # NB096148
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 215-514 of human HBP1 (NP_036389.2). |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: FLJ16340 High mobility group box transcription factor 1 HMG box transcription factor 1 HMG box-containing protein 1 HMG-box containing protein 1 HMG-box transcription factor 1 USMCC2; High mobility group box transcription factor 1; HMG box transcription factor 1; HMG box-containing protein 1
Gene Aliases: HBP1
UniProt ID: (Human) O60381
Entrez Gene ID: (Human) 26959
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.