Invitrogen
Manufacturer Code:PA548083
Catalog # PIPA548083
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Mouse myeloma cell line NS0-derived recombit mouse TREML4/TLT-4 Ser25-Val195 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: diphtheria toxin receptor; DTR DTS DTSF HEGFL; HB-EGF; heparin binding epidermal growth factor-like growth factor; Heparin-binding EGF-like growth factor; Proheparin-binding EGF-like growth factor
Gene Aliases: AW047313; Dtr; Dts; Hbegf; Hegfl
UniProt ID: (Mouse) Q06186
Entrez Gene ID: (Mouse) 15200
Molecular Function:
growth factor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.