Proteintech
Manufacturer Code:209751AP150UL
Catalog # 50-173-3184
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HB9 HLXB9 Homeobox protein HB9 HOXHB9 MNX1 SCRA1; homeobox gene HB9; homeobox HB9; Homeobox protein HB9; Motor neuron and pancreas homeobox protein 1
Gene Aliases: HB9; HLXB9; HOXHB9; MNR2; MNX1; SCRA1
UniProt ID: (Human) P50219
Entrez Gene ID: (Human) 3110, (Mouse) 15285
Molecular Function:
DNA binding protein
helix-turn-helix transcription factor
homeobox transcription factor
nucleic acid binding
transcription factor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.