Novus Biologicals
Manufacturer Code:NBP212352AF532
Catalog # NP21232A32
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | MM0324-3R44 |
Immunogen | Human HB-EGF |
Conjugate | Alexa Fluor 532 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: Diphtheria toxin receptor; diphtheria toxin receptor (heparin-binding EGF-like growth factor); diphtheria toxin receptor (heparin-binding epidermal growth factor-like growth factor); diphtheria toxin receptor (heparin-binding epidermal growth factor-like growthfactor) DTRHEGFLdiphtheria toxin receptor (heparin-binding EGF-like growth factor) DTS DTSF heparin-binding EGF-like growth factor heparin-binding epidermal growth factor proheparin-binding EGF-like growth factor; DT-R; HB-EGF; Heparin-binding EGF-like growth factor; heparin-binding epidermal growth factor; Proheparin-binding EGF-like growth factor
Gene Aliases: DTR; DTS; DTSF; HBEGF; HEGFL
UniProt ID: (Human) Q99075
Entrez Gene ID: (Human) 1839
Molecular Function:
growth factor
signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.