R&D Systems
Manufacturer Code:MAB8239SP
Catalog # MAB8239SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | 917525R |
Immunogen | <em>E. coli</em>-derived recombinant mouse HB-EGF Asp63-Leu148 Accession # Q06186 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Rat Monoclonal Antibody
Protein Aliases: Diphtheria toxin receptor; diphtheria toxin receptor (heparin-binding EGF-like growth factor); diphtheria toxin receptor (heparin-binding epidermal growth factor-like growth factor); diphtheria toxin receptor (heparin-binding epidermal growth factor-like growthfactor) Dtr DTRHEGFLdiphtheria toxin receptor (heparin-binding EGF-like growth factor) Dts DTSF HBEGF Hegfl heparin-binding EGF-like growth factor heparin-binding epidermal growth factor proheparin-binding EGF-like growth factor; DT-R; HB-EGF; Heparin-binding EGF-like growth factor; heparin-binding epidermal growth factor; Proheparin-binding EGF-like growth factor
Gene Aliases: DTR; DTS; DTSF; HBEGF; HEGFL
UniProt ID: (Human) Q99075
Entrez Gene ID: (Human) 1839
Molecular Function: growth factor signaling molecule
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.