Invitrogen
Manufacturer Code:PA572403
Catalog # PIPA572403
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HA synthase 2; HA synthase 2 hyaluronate synthase 2 hyaluronic acid synthase 2; Hyaluronan synthase 2; hyaluronan synthase homolog; Hyaluronate synthase 2; Hyaluronic acid synthase 2
Gene Aliases: HAS2
UniProt ID: (Bovine) O97711, (Human) Q92819, (Mouse) P70312, (Rat) O35776
Entrez Gene ID: (Horse) 100009708, (Bovine) 281220, (Chicken) 395594, (Human) 3037, (Mouse) 15117, (Rat) 25694
Molecular Function:
glycosyltransferase
transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.