Novus Biologicals
Manufacturer Code:NBP271739IR
Catalog # NBP271739IR
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | OTI5H2 |
Immunogen | Full length recombinant protein of human HARS2 (NP_036340) produced in HEK293T cells. |
Conjugate | DyLight 755 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. |
Mouse Monoclonal Antibody
Protein Aliases: EC 6.1.1 EC 6.1.1.21 HARSLprobable histidyl-tRNA synthetase mitochondrial HARS-related HARSRhistidine tRNA ligase 2 mitochondrial (putative) HisRS histidine translase Histidine--tRNA ligase Histidine--tRNA ligase-like histidyl-tRNA synthetase 2 mitochondrial (putative) histidyl-tRNA synthetase-like HO3histidine-tRNA ligase homolog; HARS-related; HisRS; histidine translase; histidine tRNA ligase 2, mitochondrial (putative); Histidine--tRNA ligase, mitochondrial; Histidine--tRNA ligase-like; histidine-tRNA ligase homolog; Histidyl-tRNA synthetase; histidyl-tRNA synthetase 2; histidyl-tRNA synthetase 2, mitochondrial (putative); probable histidyl-tRNA synthetase, mitochondrial
Gene Aliases: HARS2; HARSL; HARSR; HO3; PRLTS2
UniProt ID: (Human) P49590
Entrez Gene ID: (Human) 23438
Molecular Function: RNA binding protein aminoacyl-tRNA synthetase ligase nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.