Proteintech
Manufacturer Code:113011AP150UL
Catalog # 50-172-7042
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Immunoprecipitation (IP) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | HARS2 Fusion Protein Ag1827 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HARS-related; HARS2 HARSL HARSR HisRS Histidine tRNA ligase Histidine tRNA ligase like HO3; HisRS; histidine translase; histidine tRNA ligase 2, mitochondrial (putative); Histidine--tRNA ligase, mitochondrial; Histidine--tRNA ligase-like; histidine-tRNA ligase homolog; Histidyl-tRNA synthetase; histidyl-tRNA synthetase 2; histidyl-tRNA synthetase 2, mitochondrial (putative); probable histidine--tRNA ligase, mitochondrial; probable histidyl-tRNA synthetase, mitochondrial; zinc finger, matrin type 2
Gene Aliases: 4631412B19Rik; HARS2; Hars2l; HARSL; HARSR; HO3; PRLTS2; RGD1308426; Zmat2
UniProt ID: (Human) P49590, (Mouse) Q99KK9
Entrez Gene ID: (Human) 23438, (Rat) 307491, (Mouse) 70791
Molecular Function: RNA binding protein aminoacyl-tRNA synthetase ligase nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.