Abnova
Manufacturer Code:H00003035M03
Catalog # 89-022-444
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 4D4 |
Immunogen | HARS (NP_002100 1 a.a. ~ 96 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. |
Mouse monoclonal antibody raised against a partial recombinant HARS.
For Research Use Only
Protein Aliases: FLJ20491; HisRS; histidine translase; Histidine--tRNA ligase, cytoplasmic; Histidyl-tRNA synthetase; HRS
Gene Aliases: CMT2W; HARS; HARS1; HRS; USH3B
UniProt ID: (Human) P12081
Entrez Gene ID: (Human) 3035
Molecular Function: RNA binding protein aminoacyl-tRNA synthetase ligase nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.