Invitrogen
Manufacturer Code:PA557708
Catalog # PIPA557708
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human HARS |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: dead end homolog 1; HisRS; histidine translase; histidine tRNA ligase; Histidine--tRNA ligase, cytoplasmic; Histidyl-tRNA synthetase; histidyl-tRNA synthetase, cytoplasmic; HRS HisRS histidine tRNA ligase 1 cytoplasmic histidine translase histidine--tRNA ligase cytoplasmic histidine-tRNA ligase histidyl-tRNA synthetase cytoplasmic
Gene Aliases: CMT2W; Dnd1; HARS; HARS1; HRS; MMHRS; USH3B
UniProt ID: (Human) P12081, (Mouse) Q61035
Entrez Gene ID: (Human) 3035, (Mouse) 15115, (Rat) 307492
Molecular Function:
RNA binding protein
aminoacyl-tRNA synthetase
ligase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.