Invitrogen
Manufacturer Code:PA545194
Catalog # PIPA545194
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the N-terminal of human HARS |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: dead end homolog 1; HisRS; histidine translase; histidine tRNA ligase; Histidine--tRNA ligase; Histidine--tRNA ligase, cytoplasmic; Histidyl-tRNA synthetase; Histidyl-tRNA synthetase FLJ20491 HRS USH3B; histidyl-tRNA synthetase, cytoplasmic
Gene Aliases: CMT2W; Dnd1; HARS; HARS1; HARSL; HRS; im:7136719; MMHRS; USH3B; zgc:92215
UniProt ID: (Human) P12081, (Bovine) Q2KI84, (Mouse) Q61035, (Zebrafish) E9QI36
Entrez Gene ID: (Human) 3035, (Rabbit) 100351394, (Dog) 478034, (Bovine) 510937, (Horse) 100061748, (Mouse) 15115, (Rat) 307492, (Guinea pig) 100729679, (Zebrafish) 447847
Molecular Function:
RNA binding protein
aminoacyl-tRNA synthetase
ligase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.