Invitrogen
Manufacturer Code:PA5106468
Catalog # PIPA5106468
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | A synthesized peptide derived from human HARS corresponding to a region within C-terminal amino acids. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: CMT2W dead end homolog 1 Dnd1 FLJ20491 HARS HisRS histidine translase histidine tRNA ligase Histidine--tRNA ligase cytoplasmic histidyl-tRNA synthetase histidyl-tRNA synthetase cytoplasmic HRS MMHRS USH3B; dead end homolog 1; HisRS; histidine translase; histidine tRNA ligase; Histidine--tRNA ligase, cytoplasmic; Histidyl-tRNA synthetase; histidyl-tRNA synthetase, cytoplasmic
Gene Aliases: CMT2W; Dnd1; HARS; HARS1; HRS; MMHRS; USH3B
UniProt ID: (Human) P12081, (Mouse) Q61035
Entrez Gene ID: (Human) 3035, (Mouse) 15115, (Rat) 307492
Molecular Function:
RNA binding protein
aminoacyl-tRNA synthetase
ligase
nucleic acid binding
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.