Invitrogen
Manufacturer Code:PA553999
Catalog # PIPA553999
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit protein corresponding to Human HAPLN1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Cartilage link protein; cartilage linking protein 1; Cartilage-link protein; Cartilage-linking protein 1; CRTL1 Cartilage link protein cartilage linking protein 1 cartilage-link protein cartilage-linking protein 1 proteoglycan link protein; Hyaluronan and proteoglycan link protein 1; Proteoglycan link protein
Gene Aliases: CRT1; CRTL1; HAPLN1
UniProt ID: (Human) P10915
Entrez Gene ID: (Human) 1404
Molecular Function:
extracellular matrix glycoprotein
extracellular matrix protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.