Novus Biologicals
Manufacturer Code:NB11074569PECY7
Catalog # N1074569PC7
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1B6 |
Immunogen | Fusion protein from rat HAP1. [UniProt No. P54256] |
Conjugate | PE-Cyanine7 |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C in the dark. Do not freeze. |
Mouse Monoclonal Antibody
Protein Aliases: HAP-1; HAP2huntingtin-associated protein 2 hHLP1 HIP5 HLP1 HLPHAP-1 huntingtin-associated protein 1 Neuroan 1; Huntingtin-associated protein 1; huntingtin-associated protein 2; Neuroan 1
Gene Aliases: HAP1; HAP2; hHLP1; HIP5; HLP; HLP1
UniProt ID: (Human) P54257
Entrez Gene ID: (Human) 9001
Molecular Function:
membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.