Novus Biologicals
Manufacturer Code:NBP2866630.1ML
Catalog # NB034287
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | The immunogen is a synthetic peptide directed towards the middle region of human HAP1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. Aliquot and store at -20°:C long term. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HAP-1; HAP2huntingtin-associated protein 2 hHLP1 HIP5 HLP1 HLPHAP-1 huntingtin-associated protein 1 Neuroan 1; Huntingtin-associated protein 1; huntingtin-associated protein 2; Neuroan 1
Gene Aliases: HAP1; HAP2; hHLP1; HIP5; HLP; HLP1
UniProt ID: (Human) P54257
Entrez Gene ID: (Human) 9001
Molecular Function:
membrane traffic protein
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.