Invitrogen
Manufacturer Code:PA590263
Catalog # PIPA590263
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 1-351 of human HAO2 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: (S)-2-hydroxy-acid oxidase, peroxisomal; HAOX2; hydroxyacid oxidase (glycolate oxidase) 3; Hydroxyacid oxidase 2; hydroxyacid oxidase 2, kidney; Medium chain alpha-hydroxy acid oxidase; medium-chain 2-hydroxy acid oxidase HAOX3; Medium-chain L-2-hydroxy acid oxidase
Gene Aliases: AI325478; Hao-2; Hao2; Hao3; Haox2; Haox3
UniProt ID: (Mouse) Q9NYQ2
Entrez Gene ID: (Mouse) 56185
Molecular Function: dehydrogenase isomerase ligase oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.