Novus Biologicals
Manufacturer Code:NBP247139
Catalog # NBP247139
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | Peptide with sequence C-DVSEVDTRTTIQGE from the internal region of the protein sequence according to NP_057611.1. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Goat Polyclonal Antibody
Protein Aliases: (S)-2-hydroxy-acid oxidase peroxisomal Cell growth-inhibiting gene 16 protein EC 1.1.3.15 GIG16 growth-inhibiting protein 16 HAOX2glycolate oxidase hydroxyacid oxidase 2 hydroxyacid oxidase 2 (long chain) Long chain alpha-hydroxy acid oxidase Long-chain L-2-hydroxy acid oxidase; (S)-2-hydroxy-acid oxidase, peroxisomal; Cell growth-inhibiting gene 16 protein; glycolate oxidase; HAOX2; Hydroxyacid oxidase 2; hydroxyacid oxidase 2 (long chain); Long chain alpha-hydroxy acid oxidase; Long-chain L-2-hydroxy acid oxidase
Gene Aliases: GIG16; HAO2; HAOX2
UniProt ID: (Human) Q9NYQ3
Entrez Gene ID: (Human) 51179
Molecular Function: dehydrogenase isomerase ligase oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.