Invitrogen
Manufacturer Code:MA524346
Catalog # PIMA524346
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 732410 |
Immunogen | E. coli-derived recombit human Hydroxyacid Oxidase-1/HAO-1 Leu2-Ile370 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: (S)-2-hydroxy-acid oxidase; Glycolate oxidase; glycolate oxidase 1; Glycolate oxidase antibody GOX1 antibody HAOX1 antibody Hydroxyacid oxidase 1antibody MGC142225 antibody; GOX; HAOX1; hydroxyacid oxidase (glycolate oxidase) 1; Hydroxyacid oxidase 1; hydroxyacid oxidase 1, liver
Gene Aliases: GOX; GOX1; Hao-1; HAO1; HAOX1; XDH1
UniProt ID: (Human) Q9UJM8, (Mouse) Q9WU19
Entrez Gene ID: (Human) 54363, (Rat) 311446, (Mouse) 15112
Molecular Function:
dehydrogenase
isomerase
ligase
oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.