Abnova
Manufacturer Code:MAB16373
Catalog # 89-999-173
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Flow Cytometry (Flow) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Mouse |
Class | Monoclonal |
Type | Antibody |
Clone | 1G10 |
Immunogen | Recombinant protein corresponding to full length human HAND1 from <i>E. coli</i>. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Mouse Monoclonal Antibody
For Research Use Only
Protein Aliases: bHLHa27; Class A basic helix-loop-helix protein 27; eHAND; Extraembryonic tissues, heart, autonomic nervous system and neural crest derivatives-expressed protein 1; Heart- and neural crest derivatives-expressed protein 1; Hxt; Thing1
Gene Aliases: BHLHA27; EHAND; HAND1; Hxt; Thing1
UniProt ID: (Human) O96004
Entrez Gene ID: (Human) 9421
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.