Invitrogen
Manufacturer Code:PA542755
Catalog # PIPA542755
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptide directed towards the C-terminal of human HAL |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Histidase; Histidine ammonia-lyase; Histidine ammonia-lyase HIS HSTD; histidinemia
Gene Aliases: HAL; HIS; histidase; Hsd; HSTD; Huth; zgc:136980
UniProt ID: (Human) P42357, (Bovine) A7YWP4, (Rat) P21213, (Mouse) P35492
Entrez Gene ID: (Human) 3034, (Rabbit) 100357471, (Dog) 475433, (Bovine) 615205, (Horse) 100052193, (Rat) 29301, (Mouse) 15109, (Guinea pig) 100719999, (Zebrafish) 100004331
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.