Proteintech
Manufacturer Code:259401AP150UL
Catalog # 50-173-5134
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | HAL Fusion Protein Ag22985 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HAL HIS Histidase histidine ammonia lyase HSTD; Histidase; Histidine ammonia-lyase; histidinemia
Gene Aliases: HAL; HIS; histidase; Hsd; HSTD; Huth
UniProt ID: (Human) P42357, (Rat) P21213, (Mouse) P35492
Entrez Gene ID: (Human) 3034, (Rat) 29301, (Mouse) 15109
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.