Novus Biologicals
Manufacturer Code:NBP212356
Catalog # NBP212356
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Mouse |
Host / Isotype | Rat |
Class | Monoclonal |
Type | Antibody |
Clone | RM0088-3G21 |
Immunogen | Mouse HGFA Inhibitor 1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Rat Monoclonal Antibody
For Research Use Only
Protein Aliases: HAI HAI1 HAI-1 hepatocyte growth factor activator inhibitor 1 Hepatocyte growth factor activator inhibitor type 1 kunitz-type protease inhibitor 1 MANSC2 serine peptidase inhibitor Kunitz type 1 serine protease inhibitor Kunitz type 1; HAI-1; Hepatocyte growth factor activator inhibitor type 1; Kunitz-type protease inhibitor 1; serine protease inhibitor, Kunitz type 1
Gene Aliases: HAI; HAI1; MANSC2; SPINT1; UNQ223/PRO256
UniProt ID: (Human) O43278
Entrez Gene ID: (Human) 6692
Molecular Function:
enzyme modulator
protease inhibitor
serine protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.