Proteintech
Manufacturer Code:275931AP150UL
Catalog # 50-173-5710
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | HAI-1 Fusion Protein Ag26733 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: HAI HAI 1 HAI1 MANSC2 SPINT1; HAI-1; Hepatocyte growth factor activator inhibitor type 1; Kunitz-type protease inhibitor 1; serine protease inhibitor, Kunitz type 1
Gene Aliases: HAI; HAI1; MANSC2; SPINT1; UNQ223/PRO256
UniProt ID: (Human) O43278
Entrez Gene ID: (Human) 6692
Molecular Function:
enzyme modulator
protease inhibitor
serine protease inhibitor
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.