Invitrogen
Manufacturer Code:PA593097
Catalog # PIPA593097
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fusion protein containing a sequence corresponding to amino acids 1-260 of human HAGH |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | -20° C Avoid Freeze/Thaw Cycles |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: Glx II; Glyoxalase II; glyoxylase 2; Hydroxyacylglutathione hydrolase, mitochondrial; hydroxyacylglutathione hydroxylase; Round spermatid protein RSP29
Gene Aliases: BC019817; Glo-2; GLO2; GLX2; GLXII; HAGH; HAGH1; RSP29
UniProt ID: (Human) Q16775, (Mouse) Q99KB8, (Rat) O35952
Entrez Gene ID: (Human) 3029, (Mouse) 14651, (Rat) 24439
Molecular Function:
hydrolase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.