Novus Biologicals
Manufacturer Code:NBP15473220UL
Catalog # NBP15473220
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (IHC) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Synthetic peptides corresponding to HADH(hydroxyacyl-Coenzyme A dehydrogenase) The peptide sequence was selected from the C terminal of HADH. Peptide sequence YPMGPFELLDYVGLDTTKFIVDGWHEMDAENPLHQPSPSLNKLVAENKFG. |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at -20C. Avoid freeze-thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: EC 1.1.1 EC 1.1.1.35 HADH1 HADHSChydroxyacyl-Coenzyme A dehydrogenase HCDH hydroxyacyl-CoA dehydrogenase L-3-hydroxyacyl-Coenzyme A dehydrogenase short chain Medium and short-chain L-3-hydroxyacyl-coenzyme A dehydrogenase MGC8392 mitochondrial MSCHAD SCHADHHF4; HCDH; Hydroxyacyl-coenzyme A dehydrogenase, mitochondrial; L-3-hydroxyacyl-Coenzyme A dehydrogenase, short chain; Medium and short-chain L-3-hydroxyacyl-coenzyme A dehydrogenase; Short-chain 3-hydroxyacyl-CoA dehydrogenase; testis secretory sperm-binding protein Li 203a
Gene Aliases: HAD; HAD1; HADH; HADH1; HADHSC; HCDH; HHF4; MSCHAD; SCHAD
UniProt ID: (Human) Q16836
Entrez Gene ID: (Human) 3033
Molecular Function: dehydrogenase epimerase/racemase hydratase isomerase lyase oxidoreductase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.