Invitrogen
Manufacturer Code:PA528511
Catalog # PIPA528511
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunocytochemistry (ICC) | Assay dependent |
Immunofluorescence (IF) | Assay dependent |
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit fragment corresponding to a region within amino acids 337 and 578 of Human HACL1 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 1600020H07Rik; 2-HPCL; 2-HPCL HPCL HPCL2 PHYH2 1600020H07Rik 2-hydroxyphytanol-CoA lyase 2-hydroxyphytanoyl-CoA lyase phytanoyl-CoA 2-hydroxylase 2 phytanoyl-CoA hydroxylase 2; 2-hydroxyacyl-CoA lyase 1; 2-hydroxyphytanol-CoA lyase; 2-hydroxyphytanoyl-CoA lyase; Phytanoyl-CoA 2-hydroxylase 2; phytanoyl-CoA hydroxylase 2
Gene Aliases: 1600020H07Rik; 2-HPCL; HACL1; HPCL; HPCL2; HSPC279; PHYH2
UniProt ID: (Human) Q9UJ83, (Mouse) Q9QXE0
Entrez Gene ID: (Human) 26061, (Mouse) 56794
Molecular Function: decarboxylase dehydrogenase lyase oxidoreductase transferase
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.