Invitrogen
Manufacturer Code:PA571820
Catalog # PIPA571820
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Immunohistochemistry (Paraffin) (IHC (P)) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Human |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | KLH conjugated synthetic peptide between 37-66 amino acids from the N-terminal region of human PTPLA |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Store at 4°C short term. For long term storage store at -20°C avoiding freeze/thaw cycles. |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3-hydroxyacyl-CoA dehydratase 1; CAP; cementum attachment protein; Cementum-attachment protein; HACD1; protein tyrosine phosphatase-like (proline instead of catalytic arginine), member A; Protein-tyrosine phosphatase-like member A; very-long-chain (3R)-3-hydroxyacyl-[acyl-carrier protein] dehydratase 1; Very-long-chain (3R)-3-hydroxyacyl-CoA dehydratase 1; Very-long-chain (3R)-3-hydroxyacyl-CoA dehydratase 1 3-hydroxyacyl-CoA dehydratase 1 HACD1 Cementum attachment protein Protein-tyrosine phosphatase-like member A PTPLA HACD1
Gene Aliases: CAP; HACD1; PTPLA
UniProt ID: (Human) B0YJ81
Entrez Gene ID: (Human) 9200
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.