R&D Systems
Manufacturer Code:AF5359SP
Catalog # AF5359SP
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
Western Blot (WB) | Assay dependent |
Species reactivity | Many |
Host / Isotype | Goat |
Class | Polyclonal |
Type | Antibody |
Immunogen | <EM>E. coli</EM>-derived recombinant mouse HABP1/C1QBP Leu72-Gln279 Accession # NP_031599 |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt -20 to -70 degreesC as supplied. 1 month 2 to 8 degreesC under sterile conditions after reconstitution. 6 months -20 to -70 degreesC under sterile con |
Goat Polyclonal Antibody
For Research Use Only
Protein Aliases: ASF/SF2-associated protein p32; C1q globular domain-binding protein; C1q globular domain-binding protein C1qBP complement component 1 Q subcomponent-binding protein mitochondrial complement component 1 q subcomponent binding protein gC1qBP gC1qR gC1Q-R GC1q-R protein Glycoprotein gC1qBP HABP1p33 Hyaluronan-binding protein 1 Mitochondrial matrix protein p32 p32 SF2P32 splicing factor SF2-associated protein; C1qBP; Complement component 1 Q subcomponent-binding protein, mitochondrial; complement component 1, q subcomponent binding protein; gC1q-R protein; Glycoprotein gC1qBP; Hyaluronan-binding protein 1; Mitochondrial matrix protein p32; p33; SF2AP32; splicing factor SF2-associated protein
Gene Aliases: C1QBP; gC1Q-R; GC1QBP; gC1qR; HABP1; p32; SF2P32
UniProt ID: (Human) Q07021
Entrez Gene ID: (Human) 708
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.