Invitrogen
Manufacturer Code:PA581604
Catalog # PIPA581604
Quantity:
TESTED APPLICATIONS | DILUTION |
---|---|
ELISA (ELISA) | Assay dependent |
Western Blot (WB) | Assay dependent |
Species reactivity | Rat |
Host / Isotype | Rabbit |
Class | Polyclonal |
Type | Antibody |
Immunogen | Recombit Rat HAAO/3-HAO Protein |
Conjugate | Unconjugated |
Form | Liquid |
Purification | purified |
Storage buffer | PBS, pH7.4 |
Contains | 15mM sodium azide |
Storage conditions | Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C |
Rabbit Polyclonal Antibody
For Research Use Only
Protein Aliases: 3-HAO; 3-HAO HAO 3-hydroxyanthranilate oxygenase 3-hydroxyanthranilic acid dioxygenase HAD; 3-HAO {ECO:0000255|HAMAP-Rule:MF_03019}; 3-hydroxyanthranilate 3,4-dioxygenase; 3-hydroxyanthranilate 3,4-dioxygenase {ECO:0000255|HAMAP-Rule:MF_03019}; 3-hydroxyanthranilate oxygenase; 3-hydroxyanthranilate oxygenase {ECO:0000255|HAMAP-Rule:MF_03019}; 3-hydroxyanthranilic acid dioxygenase; 3-hydroxyanthranilic acid dioxygenase {ECO:0000255|HAMAP-Rule:MF_03019}; HAD; HAD {ECO:0000255|HAMAP-Rule:MF_03019}
Gene Aliases: Haao
UniProt ID: (Rat) P46953
Entrez Gene ID: (Rat) 56823
If an Invitrogen™ antibody doesn’t perform as described on our website or datasheet, we’ll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.